Product Details

SNP ID
rs61746176
Assay Type
Functionally tested
NCBI dbSNP Submissions
5
Location
Chr.1:200873524 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTGCGCGCTGGCCTCGTGCTGCGGC[G/T]TCTGGGCCGTGGCGCTGCTGGCCGG
Phenotype
MIM: 602174
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
GPR25 PubMed Links

Gene Details

Gene
GPR25
Gene Name
G protein-coupled receptor 25
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005298.3 570 Missense Mutation GTC,TTC V163F NP_005289.2

View Full Product Details