Product Details

SNP ID
rs61995862
Assay Type
Functionally tested
NCBI dbSNP Submissions
13
Location
Chr.1:156252929 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTCCAATGTACTTACCTGTCCTTGG[A/G]ATGATGACAATGAAGCGGCCACTGG
Phenotype
MIM: 112260 MIM: 614579 MIM: 610962
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
BGLAP PubMed Links

Gene Details

Gene
BGLAP
Gene Name
bone gamma-carboxyglutamate protein
There are no transcripts associated with this gene.

Gene
PAQR6
Gene Name
progestin and adipoQ receptor family member 6
There are no transcripts associated with this gene.

Gene
PMF1-BGLAP
Gene Name
PMF1-BGLAP readthrough
There are no transcripts associated with this gene.

Gene
SMG5
Gene Name
SMG5, nonsense mediated mRNA decay factor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001323614.1 2747 Silent Mutation ATC,ATT I831I NP_001310543.1
NM_001323615.1 2747 Silent Mutation ATC,ATT I838I NP_001310544.1
NM_001323616.1 2747 Silent Mutation ATC,ATT I818I NP_001310545.1
NM_001323617.1 2747 Silent Mutation ATC,ATT I818I NP_001310546.1
NM_015327.2 2747 Silent Mutation ATC,ATT I884I NP_056142.2
XM_017000842.1 2747 Silent Mutation ATC,ATT I672I XP_016856331.1
XM_017000843.1 2747 Silent Mutation ATC,ATT I623I XP_016856332.1

View Full Product Details