Product Details
- SNP ID
-
rs3111341
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.2:232457036 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- ACCAGTGCAACACGACACGCGGCAA[A/T]GAGGTCATCTCCGTGATGAACCGGG
- Phenotype
-
MIM: 171740
- Polymorphism
- A/T, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
ALPI
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs2272422] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- ALPI
- Gene Name
- alkaline phosphatase, intestinal
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001631.4 |
526 |
Silent Mutation |
AAA,AAT |
K146N |
NP_001622.2 |
View Full Product Details