Product Details

SNP ID
rs61740983
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:179333027 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TACCGAGACGTCAAATGACTTGCCA[A/G]AATTTGCAGATGGAATCAAGGCCAG
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ZNF639 PubMed Links

Gene Details

Gene
ZNF639
Gene Name
zinc finger protein 639
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001303425.1 953 Missense Mutation AAA,GAA K70E NP_001290354.1
NM_001303426.1 953 Missense Mutation AAA,GAA K70E NP_001290355.1
NM_016331.2 953 Missense Mutation AAA,GAA K70E NP_057415.1
XM_017006550.1 953 Missense Mutation AAA,GAA K70E XP_016862039.1
XM_017006551.1 953 Missense Mutation AAA,GAA K70E XP_016862040.1
XM_017006552.1 953 Missense Mutation AAA,GAA K70E XP_016862041.1
XM_017006553.1 953 Missense Mutation AAA,GAA K70E XP_016862042.1

View Full Product Details