Product Details

SNP ID
rs61735905
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:144110019 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGAGTGTGCACGATCCTGGTTCTCA[C/G]GTCCTGGTCATGTGGGTTTGCCCTG
Phenotype
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
OR2A2 PubMed Links

Gene Details

Gene
OR2A2
Gene Name
olfactory receptor family 2 subfamily A member 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001005480.2 437 Missense Mutation ACG,AGG T146R NP_001005480.2

View Full Product Details