Product Details

SNP ID
rs61737475
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:90164518 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTTCCCTTCTACTGGGCACAATACA[C/T]GCATTGATTTTTGCCTGGAATAAGT
Phenotype
MIM: 604415
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
STEAP1 PubMed Links

Gene Details

Gene
STEAP1
Gene Name
six transmembrane epithelial antigen of the prostate 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_012449.2 1004 Silent Mutation CAC,CAT H268H NP_036581.1
Gene
STEAP2-AS1
Gene Name
STEAP2 antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details