Product Details

SNP ID
rs61738484
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:5343241 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTGTATAAAGTAGGCCTGGGAAAAG[C/G]AGGCCGCACTTCCAATCTCCCTGTG
Phenotype
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
OR51B5 PubMed Links

Gene Details

Gene
OR51B5
Gene Name
olfactory receptor family 51 subfamily B member 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001005567.2 731 Missense Mutation TCC,TGC S95C NP_001005567.2
Gene
OR51B6
Gene Name
olfactory receptor family 51 subfamily B member 6
There are no transcripts associated with this gene.

View Full Product Details