Product Details

SNP ID
rs73985231
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:39934635 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAGTGAATTCTCAATAAATGTTAAC[C/T]GAACTATGTAACGGAACTGTTACCA
Phenotype
MIM: 610075
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LRRC3C PubMed Links

Gene Details

Gene
LRRC3C
Gene Name
leucine rich repeat containing 3C
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001195545.1 Intron NP_001182474.1
XM_011524158.2 Intron XP_011522460.1
XM_017024003.1 Intron XP_016879492.1
Gene
ORMDL3
Gene Name
ORMDL sphingolipid biosynthesis regulator 3
There are no transcripts associated with this gene.

View Full Product Details