Product Details

SNP ID
rs73545145
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:41677407 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCCCGAAATGCAGAAACTACACCAA[A/G]GCTGCTATATCAGAGCCATCCCAGC
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CEACAM7 PubMed Links

Gene Details

Gene
CEACAM7
Gene Name
carcinoembryonic antigen related cell adhesion molecule 7
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001291485.1 Intron NP_001278414.1
NM_006890.4 Intron NP_008821.2

View Full Product Details