Product Details

SNP ID
rs61751885
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:53258844 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGCAACTTTTGGGTTGAAACATTTT[G/T]ACAATTATTTTGGATGCAAATTTCT
Phenotype
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
VN1R2 PubMed Links

Gene Details

Gene
VN1R2
Gene Name
vomeronasal 1 receptor 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_173856.2 553 Missense Mutation GAC,TAC D157Y NP_776255.2
Gene
VN1R4
Gene Name
vomeronasal 1 receptor 4
There are no transcripts associated with this gene.

Gene
ZNF677
Gene Name
zinc finger protein 677
There are no transcripts associated with this gene.

View Full Product Details