Product Details

SNP ID
rs61736273
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:11806049 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACATGCTGAACAAGAAAACTCTTCC[C/T]GGAGTAAAATCATGTGAAAGTGGTA
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ZNF440 PubMed Links

Gene Details

Gene
ZNF440
Gene Name
zinc finger protein 440
There are no transcripts associated with this gene.

Gene
ZNF491
Gene Name
zinc finger protein 491
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_152356.3 322 Silent Mutation CCC,CCT P32P NP_689569.2
XM_005259730.3 322 Silent Mutation CCC,CCT P32P XP_005259787.1

View Full Product Details