Product Details

SNP ID
rs147154481
Assay Type
Functionally Tested
NCBI dbSNP Submissions
6
Location
Chr.20:4854409 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TAAAATGGTGTGTGTGTGTGTGTGT[A/G]TGTGTGTTACATGTTCAAGTTTATT
Phenotype
MIM: 603791
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
SLC23A2 PubMed Links

Gene Details

Gene
SLC23A2
Gene Name
solute carrier family 23 member 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005116.5 4914 UTR 3 NP_005107.4
NM_203327.1 4914 UTR 3 NP_976072.1
XM_011529414.2 4914 UTR 3 XP_011527716.1
XM_011529415.2 4914 UTR 3 XP_011527717.1
XM_011529416.2 4914 UTR 3 XP_011527718.1
XM_011529417.2 4914 UTR 3 XP_011527719.1
XM_017028171.1 4914 UTR 3 XP_016883660.1
XM_017028172.1 4914 UTR 3 XP_016883661.1
XM_017028173.1 4914 UTR 3 XP_016883662.1
XM_017028174.1 4914 UTR 3 XP_016883663.1
XM_017028175.1 4914 UTR 3 XP_016883664.1
XM_017028176.1 4914 UTR 3 XP_016883665.1
XM_017028177.1 4914 UTR 3 XP_016883666.1
XM_017028178.1 4914 UTR 3 XP_016883667.1
XM_017028179.1 4914 UTR 3 XP_016883668.1
XM_017028180.1 4914 UTR 3 XP_016883669.1
XM_017028181.1 4914 Intron XP_016883670.1

View Full Product Details