Product Details

SNP ID
rs73905443
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.21:37220738 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAGTAGAGCAACGCACAGCACATCA[C/T]ATCGCATCCTCCGTAGCGGGAGGCT
Phenotype
MIM: 605298
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
DSCR3 PubMed Links

Gene Details

Gene
DSCR3
Gene Name
DSCR3 arrestin fold containing
There are no transcripts associated with this gene.

Gene
DSCR9
Gene Name
Down syndrome critical region 9 (non-protein coding)
There are no transcripts associated with this gene.

View Full Product Details