Product Details

SNP ID
rs62637689
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.X:137566853 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCGCCGCCGCCGCCGCCGCCGCCGC[C/T]GCCTTCAAGCTGAGCCCTGCCGCGG
Phenotype
MIM: 300265
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
ZIC3 PubMed Links

Gene Details

Gene
ZIC3
Gene Name
Zic family member 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003413.3 667 Silent Mutation GCC,GCT A54A NP_003404.1
XM_017029802.1 667 Silent Mutation GCC,GCT A54A XP_016885291.1

View Full Product Details