Product Details

SNP ID
rs115040192
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:108884947 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTGGATGGTGGCAATGGCGCTAAGG[A/G]TGATGGAGATGATGGTGATGATGTT
Phenotype
MIM: 124050
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
DAO PubMed Links

Gene Details

Gene
DAO
Gene Name
D-amino acid oxidase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001917.4 Intron NP_001908.3
XM_005268692.3 Intron XP_005268749.1
XM_011538004.2 Intron XP_011536306.1
XM_011538005.2 Intron XP_011536307.1

View Full Product Details