Product Details
- SNP ID
-
rs111863112
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.3:46408372 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- ACCCTGCCCTTCTGGGCTCATGCTG[A/G]GGGCGATCCCATGTGTAAAATTCTC
- Phenotype
-
MIM: 608379
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
CCRL2
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs6808835] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- CCRL2
- Gene Name
- C-C motif chemokine receptor like 2
- Gene
- LOC102724297
- Gene Name
- uncharacterized LOC102724297
There are no transcripts associated with this gene.
View Full Product Details