Product Details

SNP ID
rs57433624
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:144106895 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGCTGGACATGGAGCTGGGGGAGGA[A/G]GAGGTGGAGGAAGAAAGCAGAAGCG
Phenotype
MIM: 123980 MIM: 610210 MIM: 611885
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CYC1 PubMed Links

Gene Details

Gene
CYC1
Gene Name
cytochrome c1
There are no transcripts associated with this gene.

Gene
MAF1
Gene Name
MAF1 homolog, negative regulator of RNA polymerase III
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_032272.4 1305 Silent Mutation GAA,GAG E227E NP_115648.2
XM_017013903.1 1305 Silent Mutation GAA,GAG E227E XP_016869392.1
XM_017013904.1 1305 Intron XP_016869393.1
Gene
SHARPIN
Gene Name
SHANK associated RH domain interactor
There are no transcripts associated with this gene.

Gene
WDR97
Gene Name
WD repeat domain 97
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001316309.1 1305 Intron NP_001303238.1
XM_017013385.1 1305 Intron XP_016868874.1
XM_017013386.1 1305 Intron XP_016868875.1
XM_017013387.1 1305 Intron XP_016868876.1
XM_017013388.1 1305 Intron XP_016868877.1
XM_017013389.1 1305 Intron XP_016868878.1
XM_017013390.1 1305 Intron XP_016868879.1
XM_017013391.1 1305 Intron XP_016868880.1
XM_017013392.1 1305 Intron XP_016868881.1
XM_017013393.1 1305 Intron XP_016868882.1
XM_017013394.1 1305 Intron XP_016868883.1

View Full Product Details