Product Details

SNP ID
rs41314459
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:137460490 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGACTCGTCCTCCTGCAAGCAGACC[A/G]CATGTTCCAGGTGAGGACCAGGCAG
Phenotype
MIM: 608137 MIM: 612122
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
NSMF PubMed Links

Gene Details

Gene
NSMF
Gene Name
NMDA receptor synaptonuclear signaling and neuronal migration factor
There are no transcripts associated with this gene.

Gene
PNPLA7
Gene Name
patatin like phospholipase domain containing 7
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001098537.2 Intron NP_001092007.2
NM_152286.4 Intron NP_689499.4
XM_006717102.1 Intron XP_006717165.1
XM_006717104.3 Intron XP_006717167.1
XM_011518664.2 Intron XP_011516966.1
XM_017014709.1 Intron XP_016870198.1

View Full Product Details