Product Details

SNP ID
rs41293076
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:35560555 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGCAGCCTACCTCTTCACATAGATG[G/T]CACTCGACATCTCTCCGGTGAGTGG
Phenotype
MIM: 616971
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
ERGIC3 PubMed Links

Gene Details

Gene
ERGIC3
Gene Name
ERGIC and golgi 3
There are no transcripts associated with this gene.

Gene
FER1L4
Gene Name
fer-1 like family member 4, pseudogene
There are no transcripts associated with this gene.

View Full Product Details