Product Details

SNP ID
rs56792221
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.5:139388252 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGGAATTCAGAGAGGGAAAGCCCCT[A/G]GCCCAAGGTCACAAGGTGACCTTGT
Phenotype
MIM: 609447 MIM: 603790
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
MZB1 PubMed Links

Gene Details

Gene
MZB1
Gene Name
marginal zone B and B1 cell specific protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_016459.3 Intron NP_057543.2
Gene
PROB1
Gene Name
proline rich basic protein 1
There are no transcripts associated with this gene.

Gene
SLC23A1
Gene Name
solute carrier family 23 member 1
There are no transcripts associated with this gene.

Gene
SPATA24
Gene Name
spermatogenesis associated 24
There are no transcripts associated with this gene.

View Full Product Details