Product Details

SNP ID
rs132630271
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:48683953 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCTCCTCCAGGACCACGAGAACCAG[C/T]GACTCTTTGAGATGCTTGGACGAAA
Phenotype
MIM: 300392
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
WAS PubMed Links

Gene Details

Gene
WAS
Gene Name
Wiskott-Aldrich syndrome
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000377.2 162 Silent Mutation CGA,TGA R34* NP_000368.1
XM_011543977.2 162 Silent Mutation CGA,TGA R34* XP_011542279.1
XM_017029786.1 162 Silent Mutation CGA,TGA R34* XP_016885275.1

View Full Product Details