Product Details

SNP ID
rs41264275
Assay Type
Functionally tested
NCBI dbSNP Submissions
34
Location
Chr.1:204037323 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAACAGTCTCGGGGCGGGGGTATGG[A/G]CAAAGGAGTGACCTCAGCTGCCAAT
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
LINC00303 PubMed Links

Gene Details

Gene
LINC00303
Gene Name
long intergenic non-protein coding RNA 303
There are no transcripts associated with this gene.

View Full Product Details