Product Details

SNP ID
rs41264229
Assay Type
Functionally tested
NCBI dbSNP Submissions
21
Location
Chr.1:155336649 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGGCTAGGGGTGAAGAGAAATATAC[C/T]GTCTCTTTAAGAAAGGGACATTGAA
Phenotype
MIM: 607999
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ASH1L PubMed Links

Gene Details

Gene
ASH1L
Gene Name
ASH1 like histone lysine methyltransferase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_018489.2 4881 UTR 3 NP_060959.2
XM_005245337.4 4881 Intron XP_005245394.1
XM_006711450.3 4881 UTR 3 XP_006711513.1
XM_006711451.3 4881 UTR 3 XP_006711514.1
XM_006711452.3 4881 UTR 3 XP_006711515.1
XM_011509769.2 4881 Intron XP_011508071.1
XM_011509770.2 4881 UTR 3 XP_011508072.1
XM_017001784.1 4881 UTR 3 XP_016857273.1
XM_017001785.1 4881 UTR 3 XP_016857274.1
XM_017001786.1 4881 Intron XP_016857275.1
XM_017001787.1 4881 Intron XP_016857276.1
XM_017001788.1 4881 UTR 3 XP_016857277.1
Gene
MIR555
Gene Name
microRNA 555
There are no transcripts associated with this gene.

Gene
RUSC1
Gene Name
RUN and SH3 domain containing 1
There are no transcripts associated with this gene.

View Full Product Details