Product Details

SNP ID
hCV86251408
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.1:156768234 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GATCGCGGCGCCGGAGTTGCATAAG[G/T]GAGATGTGAGTATCCGGGGAAGGCA
Phenotype
MIM: 179755
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
PRCC PubMed Links

Gene Details

Gene
PRCC
Gene Name
papillary renal cell carcinoma (translocation-associated)
There are no transcripts associated with this gene.

View Full Product Details