Product Details

SNP ID
rs41273227
Assay Type
Functionally tested
NCBI dbSNP Submissions
15
Location
Chr.1:156935525 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATTTACAATAGCAAAGATGCACATA[C/T]GATACATACATATTTATAGGAAACC
Phenotype
MIM: 605708
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ARHGEF11 PubMed Links

Gene Details

Gene
ARHGEF11
Gene Name
Rho guanine nucleotide exchange factor 11
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014784.3 4672 UTR 3 NP_055599.1
NM_198236.2 4672 UTR 3 NP_937879.1
XM_005245629.4 4672 UTR 3 XP_005245686.1
XM_005245633.4 4672 UTR 3 XP_005245690.1
XM_006711659.2 4672 UTR 3 XP_006711722.1
XM_006711660.3 4672 UTR 3 XP_006711723.1
XM_006711661.2 4672 UTR 3 XP_006711724.1
XM_006711662.2 4672 UTR 3 XP_006711725.1
XM_006711663.3 4672 UTR 3 XP_006711726.1
XM_006711665.3 4672 UTR 3 XP_006711728.1
XM_011510185.2 4672 UTR 3 XP_011508487.1
XM_011510186.2 4672 UTR 3 XP_011508488.1
XM_011510187.2 4672 UTR 3 XP_011508489.1
XM_011510189.2 4672 UTR 3 XP_011508491.1
XM_017002920.1 4672 UTR 3 XP_016858409.1
XM_017002921.1 4672 UTR 3 XP_016858410.1
XM_017002922.1 4672 UTR 3 XP_016858411.1
XM_017002923.1 4672 UTR 3 XP_016858412.1
XM_017002924.1 4672 UTR 3 XP_016858413.1
Gene
LRRC71
Gene Name
leucine rich repeat containing 71
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_144702.2 4672 Intron NP_653303.2
XM_005244926.3 4672 Intron XP_005244983.1
XM_005244927.3 4672 Intron XP_005244984.1
XM_005244928.2 4672 Intron XP_005244985.1
XM_006711185.3 4672 Intron XP_006711248.1
XM_006711186.3 4672 Intron XP_006711249.1
XM_006711187.3 4672 Intron XP_006711250.1
XM_011509239.2 4672 Intron XP_011507541.1
XM_011509240.2 4672 Intron XP_011507542.1
XM_011509241.2 4672 Intron XP_011507543.1
XM_017000459.1 4672 Intron XP_016855948.1
XM_017000460.1 4672 Intron XP_016855949.1
XM_017000461.1 4672 Intron XP_016855950.1
XM_017000462.1 4672 Intron XP_016855951.1
Gene
MIR765
Gene Name
microRNA 765
There are no transcripts associated with this gene.

View Full Product Details