Product Details

SNP ID
rs41268107
Assay Type
Functionally tested
NCBI dbSNP Submissions
10
Location
Chr.1:23088443 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GATGGCTGGTAAATCTTATGGGGTA[C/T]TTGAGCCCATTGGGCAAATGAGGAG
Phenotype
MIM: 609132 MIM: 601422
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
KDM1A PubMed Links

Gene Details

Gene
KDM1A
Gene Name
lysine demethylase 1A
There are no transcripts associated with this gene.

Gene
LUZP1
Gene Name
leucine zipper protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001142546.1 4067 UTR 3 NP_001136018.1
NM_033631.3 4067 UTR 3 NP_361013.3
XM_011542090.2 4067 UTR 3 XP_011540392.1
XM_011542091.2 4067 UTR 3 XP_011540393.1
XM_017002252.1 4067 UTR 3 XP_016857741.1

View Full Product Details