Product Details

SNP ID
rs41464145
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:162373675 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TTGGGATTCAAGCCTACAGAACAGA[C/T]AGAAGTAGGAAAAGACAGTCTAAAA
Phenotype
MIM: 607030 MIM: 608169
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
GCA PubMed Links

Gene Details

Gene
GCA
Gene Name
grancalcin
There are no transcripts associated with this gene.

Gene
KCNH7
Gene Name
potassium voltage-gated channel subfamily H member 7
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_033272.3 Intron NP_150375.2
NM_173162.2 Intron NP_775185.1
XM_011512109.2 Intron XP_011510411.1
XM_017005218.1 Intron XP_016860707.1
XM_017005219.1 Intron XP_016860708.1
XM_017005220.1 Intron XP_016860709.1
XM_017005221.1 Intron XP_016860710.1

View Full Product Details