Product Details

SNP ID
rs41295924
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:100627172 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCTGGATTGCCAGAGAGGACCTAGA[C/T]CCCAGGGAATGCAGAGAGAAGGGGC
Phenotype
MIM: 604720
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
TFR2 PubMed Links

Gene Details

Gene
TFR2
Gene Name
transferrin receptor 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001206855.1 Intron NP_001193784.1
NM_003227.3 Intron NP_003218.2
XM_005250553.4 Intron XP_005250610.1
XM_017012573.1 Intron XP_016868062.1

View Full Product Details