Product Details

SNP ID
rs41298202
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:35698945 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GACTCATCTGCCTCCTGCAATAAGC[A/G]AAGGTTGCAGAAAGAGATGTAAAGT
Phenotype
MIM: 186745 MIM: 190990
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
TLN1 PubMed Links

Gene Details

Gene
TLN1
Gene Name
talin 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006289.3 Intron NP_006280.3
Gene
TPM2
Gene Name
tropomyosin 2 (beta)
There are no transcripts associated with this gene.

View Full Product Details