Product Details

SNP ID
rs41277136
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:122908312 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTCATACCTGAACTTTAAAAAAAGA[A/T]TCCATTTTCCTGAAAATTTTTAGAC
Phenotype
MIM: 615231 MIM: 605976
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
RC3H2 PubMed Links

Gene Details

Gene
RC3H2
Gene Name
ring finger and CCCH-type domains 2
There are no transcripts associated with this gene.

Gene
ZBTB26
Gene Name
zinc finger and BTB domain containing 26
There are no transcripts associated with this gene.

Gene
ZBTB6
Gene Name
zinc finger and BTB domain containing 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006626.5 3848 UTR 3 NP_006617.1

View Full Product Details