Product Details

SNP ID
rs41305605
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.9:127941440 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGGACACTGTGCGGTCAGCTTCCTT[G/T]TTAAAGTGCCAGTATCGGTGGGGCA
Phenotype
MIM: 603564 MIM: 610891 MIM: 612865
Polymorphism
G/T, Transversion Substitution
Allele Nomenclature
Literature Links
DPM2 PubMed Links

Gene Details

Gene
DPM2
Gene Name
dolichyl-phosphate mannosyltransferase polypeptide 2, regulatory subunit
There are no transcripts associated with this gene.

Gene
FAM102A
Gene Name
family with sequence similarity 102 member A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001035254.2 2785 UTR 3 NP_001030331.1
NM_203305.2 2785 UTR 3 NP_976050.1
Gene
PIP5KL1
Gene Name
phosphatidylinositol-4-phosphate 5-kinase like 1
There are no transcripts associated with this gene.

View Full Product Details