Product Details

SNP ID
rs41307553
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:30015004 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGTCAGTGAGGAGTTTTTCTGTACT[G/T]TGCAGAAACATGAGGTTGAAAGTGA
Phenotype
MIM: 614398
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
KIAA1462 PubMed Links

Gene Details

Gene
KIAA1462
Gene Name
KIAA1462
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_020848.2 7163 UTR 3 NP_065899.1
XM_011519608.1 7163 UTR 3 XP_011517910.1
XM_011519609.2 7163 UTR 3 XP_011517911.1

View Full Product Details