Product Details

SNP ID
hCV86355156
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:30233584 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAACTCCTCAGGATCTGGTGTATAA[A/G]GACCCAGCCAGGCCCAAAATCCAGA
Phenotype
MIM: 136530
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
FSHB PubMed Links

Gene Details

Gene
FSHB
Gene Name
follicle stimulating hormone beta subunit
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000510.2 243 Silent Mutation AAA,AAG K58K NP_000501.1
NM_001018080.1 243 Silent Mutation AAA,AAG K58K NP_001018090.1

View Full Product Details