Product Details

SNP ID
rs41279122
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.13:98450310 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGACACATTTTATAGCAGAAATACA[C/G]AAGCTGTTTTTAAAGGAGGGAAATA
Phenotype
MIM: 602654 MIM: 604984
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
FARP1 PubMed Links

Gene Details

Gene
FARP1
Gene Name
FERM, ARH/RhoGEF and pleckstrin domain protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001001715.3 4524 Intron NP_001001715.2
NM_001286839.1 4524 Intron NP_001273768.1
NM_005766.3 4524 Intron NP_005757.1
XM_011521046.2 4524 Intron XP_011519348.1
XM_017020312.1 4524 Intron XP_016875801.1
XM_017020313.1 4524 Intron XP_016875802.1
Gene
STK24
Gene Name
serine/threonine kinase 24
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001032296.3 4524 UTR 3 NP_001027467.2
NM_001286649.1 4524 UTR 3 NP_001273578.1
NM_003576.4 4524 UTR 3 NP_003567.2
XM_005254079.4 4524 Intron XP_005254136.1
XM_017020794.1 4524 Intron XP_016876283.1
XM_017020795.1 4524 Intron XP_016876284.1

View Full Product Details