Product Details

SNP ID
rs45467499
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:29701343 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAGTGACGATAAAAAAATTACAAAA[A/G]TCACAAAGCAAAATATCTTTGAACC
Phenotype
MIM: 601009
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
TJP1 PubMed Links

Gene Details

Gene
TJP1
Gene Name
tight junction protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001301025.1 5641 UTR 3 NP_001287954.1
NM_001301026.1 5641 UTR 3 NP_001287955.1
NM_003257.4 5641 UTR 3 NP_003248.3
NM_175610.3 5641 UTR 3 NP_783297.2
XM_005254617.3 5641 UTR 3 XP_005254674.2
XM_005254618.3 5641 UTR 3 XP_005254675.1
XM_005254619.3 5641 UTR 3 XP_005254676.2
XM_005254620.3 5641 UTR 3 XP_005254677.2
XM_011521972.2 5641 UTR 3 XP_011520274.2
XM_017022521.1 5641 UTR 3 XP_016878010.1
XM_017022522.1 5641 UTR 3 XP_016878011.1
XM_017022523.1 5641 UTR 3 XP_016878012.1
XM_017022524.1 5641 UTR 3 XP_016878013.1
XM_017022525.1 5641 UTR 3 XP_016878014.1
XM_017022526.1 5641 UTR 3 XP_016878015.1
XM_017022527.1 5641 UTR 3 XP_016878016.1
XM_017022528.1 5641 UTR 3 XP_016878017.1
XM_017022529.1 5641 UTR 3 XP_016878018.1
XM_017022530.1 5641 UTR 3 XP_016878019.1
XM_017022531.1 5641 UTR 3 XP_016878020.1

View Full Product Details