Product Details

SNP ID
rs45516100
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:23607914 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGTACAGCATCACACCCACGCTGAG[A/C]GTCGTCTTAGGGTTAATCACAATGA
Phenotype
MIM: 610355
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
PALB2 PubMed Links

Gene Details

Gene
PALB2
Gene Name
partner and localizer of BRCA2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_024675.3 3592 Silent Mutation NP_078951.2
XM_011545946.2 3592 Silent Mutation XP_011544248.1
XM_011545947.2 3592 Intron XP_011544249.1
XM_011545948.2 3592 Silent Mutation XP_011544250.1
XM_017023671.1 3592 Intron XP_016879160.1
XM_017023672.1 3592 Intron XP_016879161.1
XM_017023673.1 3592 Intron XP_016879162.1

View Full Product Details