Product Details

SNP ID
rs41351844
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:38352622 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGGGCTTGGAATCGGAGGCCGAGAG[C/T]CTGGAGACTAACAGCTGCTCGGAAG
Phenotype
MIM: 614515 MIM: 608788
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
GPR179 PubMed Links

Gene Details

Gene
GPR179
Gene Name
G protein-coupled receptor 179
There are no transcripts associated with this gene.

Gene
SOCS7
Gene Name
suppressor of cytokine signaling 7
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014598.3 499 Silent Mutation AGC,AGT S126S NP_055413.1
XM_017024551.1 499 Silent Mutation AGC,AGT S190S XP_016880040.1
XM_017024552.1 499 Silent Mutation AGC,AGT S190S XP_016880041.1

View Full Product Details