Product Details

SNP ID
rs41339751
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:34320279 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAAAATTTCTTTCAGATTCAGTTTC[C/T]ATTCCAATCACCTGCTGCTTTAACG
Phenotype
MIM: 602283
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CCL8 PubMed Links

Gene Details

Gene
CCL8
Gene Name
C-C motif chemokine ligand 8
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005623.2 542 Silent Mutation TCC,TCT S29S NP_005614.2

View Full Product Details