Product Details

SNP ID
rs45564734
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:44003029 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCGTCTGGCGGGTCCTTCTGAACTT[A/G]TCACCTTCGCTTGGGGTCGCAACGA
Phenotype
MIM: 604086
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
LOC101928063 PubMed Links

Gene Details

Gene
LOC101928063
Gene Name
uncharacterized LOC101928063
There are no transcripts associated with this gene.

Gene
ZNF155
Gene Name
zinc finger protein 155
There are no transcripts associated with this gene.

Gene
ZNF230
Gene Name
zinc finger protein 230
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006300.3 105 UTR 5 NP_006291.2

View Full Product Details