Product Details

SNP ID
rs45540435
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:44287654 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATCACACTTGAATGGTTTTTCCCCA[G/T]TGTGGACGCTCTGATGGGCTTGAAG
Phenotype
MIM: 604749
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
ZNF235 PubMed Links

Gene Details

Gene
ZNF235
Gene Name
zinc finger protein 235
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004234.4 5940 Missense Mutation AAT,ACT N594T NP_004225.3
XM_005259412.4 5940 Missense Mutation AAT,ACT N590T XP_005259469.1

View Full Product Details