Product Details

SNP ID
rs45605232
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:44672678 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGATTCCAGAGCAGCCTCAAAAGAA[C/G]CAGGACCTTCTCCTGTCAGTCCAGG
Phenotype
MIM: 606691 MIM: 173850
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
CEACAM19 PubMed Links

Gene Details

Gene
CEACAM19
Gene Name
carcinoembryonic antigen related cell adhesion molecule 19
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001127893.1 618 Missense Mutation AAC,AAG N46K NP_001121365.1
NM_020219.3 618 Missense Mutation AAC,AAG N46K NP_064604.2
Gene
PVR
Gene Name
poliovirus receptor
There are no transcripts associated with this gene.

View Full Product Details