Product Details

SNP ID
rs41305215
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:30828812 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCATCCGATTTATATCCTACCACTT[A/C]CATGCCAATTGTCTGTACATACCAA
Phenotype
MIM: 300480
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
TAB3 PubMed Links

Gene Details

Gene
TAB3
Gene Name
TGF-beta activated kinase 1/MAP3K7 binding protein 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_152787.4 5330 UTR 3 NP_690000.3
XM_005274483.3 5330 UTR 3 XP_005274540.2
XM_005274484.2 5330 UTR 3 XP_005274541.2
XM_011545488.2 5330 Intron XP_011543790.1
XM_017029399.1 5330 Intron XP_016884888.1
XM_017029400.1 5330 Intron XP_016884889.1
XM_017029401.1 5330 Intron XP_016884890.1
XM_017029402.1 5330 Intron XP_016884891.1

View Full Product Details