Product Details
- SNP ID
-
rs41300914
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.X:126820800 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AGCCCTGGGGTCAACCTCAAGTACC[A/G]CTGAGACCTTCCGTCAATGTGCTGA
- Phenotype
-
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
PRR32
PubMed Links
Gene Details
- Gene
- PRR32
- Gene Name
- proline rich 32
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001122716.1 |
242 |
Silent Mutation |
CCA,CCG |
P54P |
NP_001116188.1 |
View Full Product Details