Product Details

SNP ID
rs41276620
Assay Type
Functionally tested
NCBI dbSNP Submissions
16
Location
Chr.1:119619779 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACTTCTCATTTTCTCCTTCCAACTC[C/G]GAGCTCAAGATTTAATCAAAACCAT
Phenotype
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
ZNF697 PubMed Links

Gene Details

Gene
ZNF697
Gene Name
zinc finger protein 697
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001080470.1 7683 UTR 3 NP_001073939.1
XM_005271315.3 7683 UTR 3 XP_005271372.1
XM_011542416.2 7683 UTR 3 XP_011540718.1
XM_017002783.1 7683 UTR 3 XP_016858272.1

View Full Product Details