Product Details
- SNP ID
-
rs41285089
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.3:33390334 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- ACAGTTCAGTCTATATAAGACATCA[C/T]TTCAAAATTATGTGGATGCCATCAC
- Phenotype
-
MIM: 605652
MIM: 609784
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
FBXL2
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs144387025] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- FBXL2
- Gene Name
- F-box and leucine rich repeat protein 2
- Gene
- UBP1
- Gene Name
- upstream binding protein 1 (LBP-1a)
View Full Product Details