Product Details

SNP ID
rs45519639
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:8879818 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATTTGCTGAATGAACTCATGTGCCT[A/G]CTTTCCCAATCTTGGGTCTCACCGC
Phenotype
MIM: 605256
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
RAD18 PubMed Links

Gene Details

Gene
RAD18
Gene Name
RAD18, E3 ubiquitin protein ligase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_020165.3 3117 UTR 3 NP_064550.3
XM_017006873.1 3117 Intron XP_016862362.1

View Full Product Details