Product Details

SNP ID
rs41311296
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:35559990 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCAGAAAGCAAAGGTTTCACACCTC[A/G]GGTCCAAAATCCCAAGATTCAAGGT
Phenotype
MIM: 616971
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ERGIC3 PubMed Links

Gene Details

Gene
ERGIC3
Gene Name
ERGIC and golgi 3
There are no transcripts associated with this gene.

Gene
FER1L4
Gene Name
fer-1 like family member 4, pseudogene
There are no transcripts associated with this gene.

View Full Product Details