Product Details

SNP ID
rs41284398
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:118005464 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTAAAGTTCTACATCTGTGCAAATG[A/G]GGGCTTTCCATGTGTGCTGACAGAC
Phenotype
MIM: 608599
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
RAB11FIP2 PubMed Links

Gene Details

Gene
RAB11FIP2
Gene Name
RAB11 family interacting protein 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014904.2 5513 UTR 3 NP_055719.1
XM_005269629.3 5513 UTR 3 XP_005269686.1
XM_017015926.1 5513 Intron XP_016871415.1

View Full Product Details