Product Details

SNP ID
rs45553933
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:18391249 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAGGGTGATACGGAATTCCAGTGAG[A/G]TCACTTCCCTTGCAGACTTTGGAAG
Phenotype
MIM: 612182
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
NAT2 PubMed Links

Gene Details

Gene
NAT2
Gene Name
N-acetyltransferase 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000015.2 5 UTR 5 NP_000006.2
XM_017012938.1 5 Intron XP_016868427.1

View Full Product Details