Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001168247.2 | 1719 | UTR 3 | NP_001161719.1 | ||
NM_001300859.1 | 1719 | UTR 3 | NP_001287788.1 | ||
NM_144990.3 | 1719 | UTR 3 | NP_659427.3 | ||
XM_005270597.2 | 1719 | UTR 3 | XP_005270654.1 | ||
XM_005270598.2 | 1719 | UTR 3 | XP_005270655.1 | ||
XM_005270599.2 | 1719 | UTR 3 | XP_005270656.1 | ||
XM_006710432.3 | 1719 | UTR 3 | XP_006710495.1 | ||
XM_011540943.1 | 1719 | UTR 3 | XP_011539245.1 | ||
XM_011540945.1 | 1719 | UTR 3 | XP_011539247.1 | ||
XM_011540947.1 | 1719 | UTR 3 | XP_011539249.1 | ||
XM_011540948.1 | 1719 | UTR 3 | XP_011539250.1 | ||
XM_011540949.1 | 1719 | UTR 3 | XP_011539251.1 | ||
XM_011540950.1 | 1719 | UTR 3 | XP_011539252.1 | ||
XM_011540951.1 | 1719 | UTR 3 | XP_011539253.1 | ||
XM_011540952.2 | 1719 | UTR 3 | XP_011539254.1 | ||
XM_011540953.2 | 1719 | UTR 3 | XP_011539255.1 | ||
XM_011540954.1 | 1719 | UTR 3 | XP_011539256.1 | ||
XM_017000573.1 | 1719 | UTR 3 | XP_016856062.1 | ||
XM_017000574.1 | 1719 | UTR 3 | XP_016856063.1 |